Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSPRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017688.2 | NP_060158.2 | 4 - 5 | 605 | 62 | |
GenBank mRNA | AJ276691.1 | - | 4 - 5 | 586 | 62 | |
AK000157.1 | - | 4 - 5 | 331 | 62 | ||
AK092607.1 | - | 4 - 5 | 596 | 62 | ||
BX462607.2 | - | 4 - 5 | 604 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |