Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006974.hg.1 |
ENST00000370487.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006974.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20006974.hg.1 |
TC20000497.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155889.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155889.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000155889.hg.1 |
TC2000008029.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000155889.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000155889.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
ENST00000370487.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008029.hg.1 |
NM_018270.5 |
Refseq |