Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301060.1 | NP_001287989.1 | 2 - 3 | 370 | 125 | |
NM_024848.2 | NP_079124.1 | 2 - 3 | 370 | 125 | ||
XM_011542169.1 | XP_011540471.1 | 2 - 3 | 370 | 125 | ||
XM_011542170.2 | XP_011540472.1 | 2 - 3 | 370 | 125 | ||
XM_011542173.2 | XP_011540475.1 | 2 - 3 | 370 | 125 | ||
XM_011542174.2 | XP_011540476.1 | 2 - 3 | 370 | 125 | ||
XM_011542176.1 | XP_011540478.1 | 2 - 3 | 370 | 125 | ||
XM_011542177.1 | XP_011540479.1 | 2 - 3 | 370 | 125 | ||
XM_017002365.1 | XP_016857854.1 | 2 - 3 | 370 | 125 | ||
GenBank mRNA | AK024003.1 | - | 2 - 3 | 173 | 125 | |
BC021704.2 | - | 2 - 3 | 326 | 4825983 | 125 | |
DB041436.1 | - | 2 - 3 | 370 | 125 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |