Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006005.hg.1 |
ENST00000299430.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006005.hg.1 |
ENST00000327305.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006005.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006005.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18006005.hg.1 |
TC18000571.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000397929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000397929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000397929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000397929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000397929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000397929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000397929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000577184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000577184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000577184.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000579169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000579169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000579169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000579169.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000579730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000579730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000579730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000579730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000579730.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000582281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000582281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
ENST00000584727.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138999.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800009011.hg.1 |
NM_138999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800161841.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161841.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161841.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800161841.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800161841.hg.1 |
TC1800009011.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800161841.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161841.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800161841.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800161841.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000327305.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000397929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000397929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000397929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000397929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000397929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000397929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000397929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000577184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000577184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000577184.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000579169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000579169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000579169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000579169.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000579730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000579730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000579730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000579730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000579730.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000582281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000582281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000583169.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
ENST00000584727.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_001201465.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138966.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138999.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800009011.hg.1 |
NM_138999.1 |
Refseq |