Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001303110.1 | NP_001290039.1 | 18 - 19 | 1135 | 92 | |
NM_001845.5 | NP_001836.3 | 18 - 19 | 1135 | 92 | ||
XM_011521048.2 | XP_011519350.1 | 18 - 19 | 9067 | 92 | ||
GenBank mRNA | AB209646.1 | - | 18 - 19 | 1120 | 92 | |
BC142626.1 | - | 18 - 19 | 1109 | 92 | ||
BC151220.1 | - | 18 - 19 | 1119 | 40148649 | 92 | |
Y00706.1 | - | 16 - 17 | 870 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |