Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003723.hg.1 |
NR_033312.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003723.hg.1 |
NR_033313.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003723.hg.1 |
NR_033315.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11003723.hg.1 |
TC11000280.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
ENST00000499568.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
ENST00000500662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
ENST00000500662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
ENST00000502161.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
ENST00000502161.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
NR_033312.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
NR_033313.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
NR_033315.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
ENST00000499568.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
ENST00000500662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
ENST00000500662.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
ENST00000502161.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
ENST00000502161.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
NR_033312.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
NR_033313.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
NR_033315.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100150240.hg.1 |
TC1100007123.hg.1 |
Clariom D TC |