Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001648.hg.1 |
ENST00000269195.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001648.hg.1 |
ENST00000537549.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001648.hg.1 |
ENST00000537748.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001648.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18001648.hg.1 |
TC18000145.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000586725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000586725.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000591081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000591081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000591081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000591081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000591924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000591924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000591924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000591924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
ENST00000591924.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800148555.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800148555.hg.1 |
TC1800007080.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800148555.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800148559.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800148559.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800148559.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800148559.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800148559.hg.1 |
TC1800007080.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800148559.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800148559.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800148559.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800148559.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000269195.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000586725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000586725.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000589189.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000590654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000591081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000591081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000591081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000591081.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000591924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000591924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000591924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000591924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
ENST00000591924.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007080.hg.1 |
NM_020474.3 |
Refseq |