Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286124.1 | NP_001273053.1 | 5 - 6 | 1063 | 185 | |
NM_001286126.1 | NP_001273055.1 | 5 - 6 | 1063 | 185 | ||
NM_018650.4 | NP_061120.3 | 5 - 6 | 1063 | 185 | ||
XM_005273134.4 | XP_005273191.1 | 5 - 6 | 930 | 185 | ||
XM_011509561.2 | XP_011507863.1 | 5 - 6 | 1059 | 185 | ||
XM_011509562.2 | XP_011507864.1 | 5 - 6 | 676 | 213 | ||
XM_017001305.1 | XP_016856794.1 | 6 - 7 | 520 | 185 | ||
XM_017001306.1 | XP_016856795.1 | 5 - 6 | 560 | 213 | ||
XM_017001307.1 | XP_016856796.1 | 6 - 7 | 960 | 185 | ||
GenBank mRNA | AF154845.1 | - | 5 - 6 | 689 | 185 | |
AK295506.1 | - | 5 - 6 | 561 | 185 | ||
AK314614.1 | - | 5 - 6 | 723 | 185 | ||
BC113869.1 | - | 5 - 6 | 599 | 40036731 | 185 | |
BC114478.1 | - | 5 - 6 | 599 | 40036730 | 185 | |
KJ534879.1 | - | 5 - 6 | 423 | 185 | ||
KJ534881.1 | - | 5 - 6 | 423 | 185 | ||
KJ535089.1 | - | 5 - 6 | 422 | 185 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Homo sapiens Probe spans exons |