Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009631.hg.1 |
ENST00000305921.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009631.hg.1 |
ENST00000392995.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009631.hg.1 |
ENST00000544712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009631.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009631.hg.1 |
TC17000739.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009625.hg.1 |
ENST00000305921.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009625.hg.1 |
ENST00000544712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009625.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009625.hg.1 |
TC17000739.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009629.hg.1 |
ENST00000305921.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009629.hg.1 |
ENST00000392995.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009629.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009629.hg.1 |
TC17000739.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000590418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000590418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000590418.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009630.hg.1 |
ENST00000305921.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009630.hg.1 |
ENST00000392995.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009630.hg.1 |
ENST00000544712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009630.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009630.hg.1 |
TC17000739.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009628.hg.1 |
ENST00000305921.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009628.hg.1 |
ENST00000392995.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009628.hg.1 |
ENST00000544712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009628.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17009628.hg.1 |
TC17000739.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000590418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000590418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000590418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700008498.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700163324.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163324.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163324.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163324.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700163324.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163324.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163324.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163324.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700163324.hg.1 |
TC1700008498.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700163322.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163322.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163322.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700163322.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163322.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163322.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700163322.hg.1 |
TC1700008498.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700163326.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163326.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163326.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163326.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700163326.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163326.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163326.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163326.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700163326.hg.1 |
TC1700008498.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700163325.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163325.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163325.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163325.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700163325.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163325.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163325.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163325.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700163325.hg.1 |
TC1700008498.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700163328.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163328.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163328.hg.1 |
ENST00000590418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163328.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163328.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700163328.hg.1 |
ENST00000305921.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163328.hg.1 |
ENST00000392995.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163328.hg.1 |
ENST00000590418.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163328.hg.1 |
ENST00000629650.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700163328.hg.1 |
NM_003620.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700163328.hg.1 |
TC1700008498.hg.1 |
Clariom D TC |