Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032525.hg.1 |
TC01002117.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032525.hg.1 |
ENST00000288774.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01032525.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000502666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000502666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000502666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000502666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000502666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000502666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000508384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000508384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000508384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000508384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000508384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000508384.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000514502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000514502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000514502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000514502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000514502.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000515760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000515760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000515760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
ENST00000515760.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000447513.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000502666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000502666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000502666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000502666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000502666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000502666.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000507596.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000508384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000508384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000508384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000508384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000508384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000508384.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000510434.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000514502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000514502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000514502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000514502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000514502.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000515760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000515760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000515760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
ENST00000515760.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_002617.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100012529.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100192674.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100192674.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100192674.hg.1 |
ENST00000288774.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100192674.hg.1 |
NM_153818.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100192674.hg.1 |
TC0100012529.hg.1 |
Clariom D TC |