Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000123.hg.1 |
ENST00000324015.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000123.hg.1 |
ENST00000382061.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000123.hg.1 |
ENST00000450523.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000123.hg.1 |
NM_000934.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000123.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000123.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR17000123.hg.1 |
TC17000014.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700144509.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700144509.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700144509.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700144509.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700144509.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700144509.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700144509.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700144509.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700144509.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700144509.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700144509.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700144509.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700144509.hg.1 |
TC1700006523.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000324015.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000382061.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000450523.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453066.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
ENST00000453723.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165920.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700006523.hg.1 |
NM_001165921.1 |
Refseq |