Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000466248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
ENST00000466248.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000482.hg.1 |
TC0X000036.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000482.hg.1 |
ENST00000380861.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000482.hg.1 |
ENST00000398613.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000482.hg.1 |
ENST00000454666.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000482.hg.1 |
ENST00000543293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X000482.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0Y00006522.hg.1 |
ENST00000610687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0Y00006522.hg.1 |
ENST00000610687.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000466248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
ENST00000466248.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00006590.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145024.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145024.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145024.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145024.hg.1 |
ENST00000380861.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145024.hg.1 |
ENST00000454666.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00145024.hg.1 |
NM_015691.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00145024.hg.1 |
TC0X00006590.hg.1 |
Clariom D TC |