Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000491605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000491605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000491605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000491605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
ENST00000491605.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000429350.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000432568.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000453743.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
ENST00000624945.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008418.hg.1 |
NR_110541.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000336726.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000403975.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000450612.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000491605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000491605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000491605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000491605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
ENST00000491605.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007043.hg.1 |
NM_152510.3 |
Refseq |