Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016358.hg.1 |
TC0X001215.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016358.hg.1 |
ENST00000324762.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016358.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016358.hg.1 |
ENST00000372905.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016358.hg.1 |
ENST00000372907.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016358.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X016358.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170341.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170341.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170341.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170341.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170341.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170341.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170341.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170341.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00170341.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170341.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00170341.hg.1 |
TC0X00010351.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000324762.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000356784.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
ENST00000372907.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_001168474.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00010351.hg.1 |
NM_024885.3 |
Refseq |