Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004827.hg.1 |
TC08000329.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004827.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004827.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004823.hg.1 |
TC08000329.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004823.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004823.hg.1 |
ENST00000531610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004823.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004826.hg.1 |
TC08000329.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004826.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004826.hg.1 |
ENST00000531610.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08004826.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151580.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151580.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151580.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151580.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151580.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151580.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151580.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151580.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151580.hg.1 |
TC0800007449.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800151581.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151581.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151581.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151581.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151581.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151581.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151581.hg.1 |
TC0800007449.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007449.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151576.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151576.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151576.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151576.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151576.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151576.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151576.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151576.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151576.hg.1 |
TC0800007449.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800151577.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151577.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151577.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151577.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151577.hg.1 |
ENST00000289957.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151577.hg.1 |
ENST00000531610.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151577.hg.1 |
ENST00000534391.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800151577.hg.1 |
NM_000749.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800151577.hg.1 |
TC0800007449.hg.1 |
Clariom D TC |