Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022052.hg.1 |
TC09001726.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022052.hg.1 |
ENST00000358701.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022052.hg.1 |
ENST00000389471.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09022052.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000455222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000455222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000455222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000455222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000455222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000455222.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181865.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181865.hg.1 |
ENST00000455222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181865.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181865.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181865.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181865.hg.1 |
ENST00000455222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181865.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900181865.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900181865.hg.1 |
TC0900011970.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000358701.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000455222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000455222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000455222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000455222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000455222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000455222.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
ENST00000616829.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011970.hg.1 |
NM_181701.3 |
Refseq |