Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012105.4 | NP_036237.2 | 8 - 9 | 90 | ||
NM_138991.2 | NP_620476.1 | 7 - 8 | 90 | |||
XM_017028314.1 | XP_016883803.1 | 8 - 9 | 90 | |||
GenBank mRNA | AF050171.1 | - | 8 - 9 | 90 | ||
AF117892.1 | - | 8 - 9 | 90 | |||
AF178532.1 | - | 8 - 9 | 1764 | 90 | ||
AF188276.1 | - | 7 - 8 | 1614 | 90 | ||
AF200192.1 | - | 8 - 9 | 90 | |||
AF200342.1 | - | 8 - 9 | 90 | |||
AF204944.1 | - | 8 - 9 | 90 | |||
AF212252.1 | - | 8 - 9 | 90 | |||
AK075539.1 | - | 9 - 10 | 90 | |||
AK292056.1 | - | 8 - 9 | 90 | |||
AY358927.1 | - | 8 - 9 | 90 | |||
BC014453.1 | - | 8 - 9 | 4868925 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Homo sapiens Probe spans exons |