Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010332.hg.1 |
ENST00000517287.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000216602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000216602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000216602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000216602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000216602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000216602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000216602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000216602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000216602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000216602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000216602.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000311141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000311141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000311141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000311141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000311141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000311141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000311141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000311141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000311141.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553512.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553512.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553512.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553512.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553512.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000553724.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554255.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554630.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000554757.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555163.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000555501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000556610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000556610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000556610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000556610.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000556795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000556795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000556795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000556795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000556795.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000602552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000602552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
ENST00000602552.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_001198953.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_001198953.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_001198953.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_001198953.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_001198953.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_001198953.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_001198953.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_001198953.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_001198953.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_001198953.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_001198953.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_024071.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_024071.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_024071.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_024071.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_024071.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_024071.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_024071.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_024071.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_024071.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400008415.hg.1 |
NM_024071.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010332.hg.1 |
ENST00000517287.1 |
Ensembl |