Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRHR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145146.1 | NP_001138618.1 | 6 - 7 | 790 | 63 | |
GenBank mRNA | HM852949.1 | - | 6 - 7 | 760 | 63 | |
L23333.1 | - | 6 - 7 | 525 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Detect Genomic DNA Homo sapiens Probe spans exons |