Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023055.hg.1 |
TC06001896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023055.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023055.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023058.hg.1 |
TC06001896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023058.hg.1 |
ENST00000275036.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023058.hg.1 |
ENST00000344726.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023058.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023058.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023058.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023058.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023056.hg.1 |
TC06001896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023056.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023054.hg.1 |
TC06001896.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023054.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185225.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185225.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185225.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185225.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185225.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185225.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185225.hg.1 |
TC0600012363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012363.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185224.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185224.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185224.hg.1 |
TC0600012363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600185226.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185226.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185226.hg.1 |
TC0600012363.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
ENST00000275036.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
ENST00000344726.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
ENST00000620514.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
NM_001201362.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
NM_001201363.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
NM_004242.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
NM_138730.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600185228.hg.1 |
TC0600012363.hg.1 |
Clariom D TC |