Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017005526.1 | XP_016861015.1 | 5 - 6 | 544 | 78 | |
GenBank mRNA | AK225873.1 | - | 5 - 6 | 495 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |