Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005247.hg.1 |
TC05000375.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005247.hg.1 |
ENST00000274368.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005247.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05005247.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000503763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000503763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000503763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000503763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000512446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000512446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000512446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000512446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
ENST00000512446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000503763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000503763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000503763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000503763.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000512446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000512446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000512446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000512446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
ENST00000512446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500013193.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193389.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193389.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193389.hg.1 |
ENST00000512446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193389.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193389.hg.1 |
ENST00000274368.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193389.hg.1 |
ENST00000506501.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193389.hg.1 |
ENST00000512446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500193389.hg.1 |
NM_001882.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500193389.hg.1 |
TC0500013193.hg.1 |
Clariom D TC |