Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010600.hg.1 |
TC20001746.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010600.hg.1 |
ENST00000374491.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010600.hg.1 |
ENST00000374492.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010600.hg.1 |
ENST00000541621.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010600.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010600.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010600.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162390.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162390.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162390.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162390.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162390.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162390.hg.1 |
TC2000008960.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000162390.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162390.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000162390.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162390.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000162390.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374491.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
ENST00000374492.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_001145025.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NM_018217.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008960.hg.1 |
NR_026728.1 |
Refseq |