Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022066.3 | NP_071349.3 | 12 - 13 | 2093 | 99 | |
XM_017024935.1 | XP_016880424.1 | 12 - 13 | 2002 | 99 | ||
GenBank mRNA | AB051521.1 | - | 12 - 13 | 2093 | 99 | |
AL832432.1 | - | 5 - 6 | 1018 | 99 | ||
BC036820.1 | - | 7 - 8 | 1229 | 99 | ||
BC051868.2 | - | 4 - 5 | 436 | 6162673 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |