Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000467683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000467683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000467683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000467683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000467683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000467683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000467683.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000474181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000474181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000474181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000474181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000474181.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000476612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000476612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000476612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000476612.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000491572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000491572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000491572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000491572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000491572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01050792.hg.1 |
TC01003293.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01050792.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01050792.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100219823.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100219823.hg.1 |
ENST00000474181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100219823.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100219823.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100219823.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100219823.hg.1 |
ENST00000474181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100219823.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100219823.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100219823.hg.1 |
TC0100015918.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000292211.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000467683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000467683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000467683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000467683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000467683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000467683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000467683.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000474181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000474181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000474181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000474181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000474181.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000476612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000476612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000476612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000476612.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000483639.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000491572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000491572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000491572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000491572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000491572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
ENST00000497453.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015918.hg.1 |
NM_017582.6 |
Refseq |