Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003696.1 | NP_001003696.1 | 3 - 4 | 632 | 191 | |
NM_001003697.1 | NP_001003697.1 | 3 - 4 | 460 | 191 | ||
NM_001003701.1 | NP_001003701.1 | 3 - 4 | 1008 | 191 | ||
NM_001003703.1 | NP_001003703.1 | 3 - 4 | 1094 | 191 | ||
NM_001320266.1 | NP_001307195.1 | 4 - 5 | 1137 | 191 | ||
NM_001320267.1 | NP_001307196.1 | 3 - 4 | 1614 | 191 | ||
NM_001685.4 | NP_001676.2 | 3 - 4 | 958 | 191 | ||
GenBank mRNA | AI085837.1 | - | 1 - 2 | 191 | ||
AL110183.1 | - | 3 - 4 | 366 | 191 | ||
BG938438.1 | - | 3 - 4 | 453 | 191 | ||
M37104.1 | - | 2 - 3 | 290 | 191 | ||
M73031.1 | - | 2 - 3 | 286 | 190 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Detect Genomic DNA Homo sapiens Probe spans exons |