Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012722.hg.1 |
TC06001074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012722.hg.1 |
ENST00000367467.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012722.hg.1 |
ENST00000537769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012722.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012723.hg.1 |
TC06001074.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012723.hg.1 |
ENST00000367467.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012723.hg.1 |
ENST00000537769.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06012723.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000470750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000470750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000470750.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000367469.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000470750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000470750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000470750.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600009772.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600166984.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600166984.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600166984.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600166984.hg.1 |
ENST00000367467.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600166984.hg.1 |
ENST00000622663.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600166984.hg.1 |
NM_015278.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600166984.hg.1 |
TC0600009772.hg.1 |
Clariom D TC |