Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000356667.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000356667.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000356667.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000356667.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000415707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000415707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000415707.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020523.hg.1 |
TC06001722.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020523.hg.1 |
ENST00000373025.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020523.hg.1 |
ENST00000424183.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06020523.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000356667.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000356667.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000356667.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000356667.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000415707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000415707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000415707.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
ENST00000425343.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_001166424.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600011800.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181007.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181007.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181007.hg.1 |
ENST00000373025.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600181007.hg.1 |
NM_002630.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600181007.hg.1 |
TC0600011800.hg.1 |
Clariom D TC |