Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321833.1 | NP_001308762.1 | 16 - 17 | 2321 | 132 | |
NM_001321834.1 | NP_001308763.1 | 16 - 17 | 3229 | 132 | ||
NM_001321835.1 | NP_001308764.1 | 16 - 17 | 1964 | 132 | ||
NM_001321836.1 | NP_001308765.1 | 17 - 18 | 2411 | 132 | ||
NM_016207.3 | NP_057291.1 | 16 - 17 | 2092 | 132 | ||
GenBank mRNA | AF171877.1 | - | 16 - 17 | 1870 | 132 | |
AK025440.1 | - | 14 - 15 | 1751 | 132 | ||
AK223487.1 | - | 16 - 17 | 1890 | 132 | ||
AK298918.1 | - | 14 - 15 | 1685 | 132 | ||
BC011654.2 | - | 16 - 17 | 1965 | 4304135 | 132 | |
BC014106.1 | - | 16 - 17 | 2882 | 132 | ||
BC020211.1 | - | 16 - 17 | 1890 | 4890593 | 132 | |
EF036508.1 | - | 3 - 4 | 252 | 132 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |