Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057267.hg.1 |
TC01003729.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057267.hg.1 |
ENST00000272203.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057267.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057267.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000485632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000485632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
ENST00000485632.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000485632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000485632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
ENST00000485632.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018553.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244509.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244509.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244509.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244509.hg.1 |
ENST00000272203.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244509.hg.1 |
ENST00000414478.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100244509.hg.1 |
NM_014935.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100244509.hg.1 |
TC0100018553.hg.1 |
Clariom D TC |