Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005862.hg.1 |
TC01000379.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005862.hg.1 |
ENST00000373836.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005862.hg.1 |
ENST00000373838.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005862.hg.1 |
ENST00000373839.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005862.hg.1 |
ENST00000493669.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005862.hg.1 |
NM_001048183.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01005862.hg.1 |
NM_023923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152913.hg.1 |
ENST00000373836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152913.hg.1 |
ENST00000373839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152913.hg.1 |
ENST00000493669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152913.hg.1 |
ENST00000632421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152913.hg.1 |
NM_001048183.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152913.hg.1 |
NM_023923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152913.hg.1 |
ENST00000373836.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152913.hg.1 |
ENST00000373839.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152913.hg.1 |
ENST00000493669.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152913.hg.1 |
ENST00000632421.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100152913.hg.1 |
NM_001048183.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152913.hg.1 |
NM_023923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100152913.hg.1 |
TC0100007556.hg.1 |
Clariom D TC |