Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002833.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002833.hg.1 |
ENST00000375454.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002833.hg.1 |
ENST00000490499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002833.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002833.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20002833.hg.1 |
TC20000227.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148971.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148971.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148971.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148971.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148971.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148971.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148971.hg.1 |
TC2000007148.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000148971.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148971.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148971.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148971.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000148971.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000148971.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375452.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000375454.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000471233.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
ENST00000490499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_001042439.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000007148.hg.1 |
NM_178466.4 |
Refseq |