Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002001.hg.1 |
TC06000190.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002001.hg.1 |
ENST00000244513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002001.hg.1 |
ENST00000377586.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002001.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002000.hg.1 |
TC06000190.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002000.hg.1 |
ENST00000244513.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002000.hg.1 |
ENST00000377586.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06002000.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600007309.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149667.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149667.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149667.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149667.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149667.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149667.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149667.hg.1 |
TC0600007309.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600149668.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149668.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149668.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149668.hg.1 |
ENST00000244513.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149668.hg.1 |
ENST00000613186.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600149668.hg.1 |
NM_001732.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600149668.hg.1 |
TC0600007309.hg.1 |
Clariom D TC |