Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000428527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000428527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000428527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000428527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000428527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000428527.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000433416.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000433416.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000433416.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000433416.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000433416.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000433416.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000438654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000438654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000438654.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000448773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000448773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000448773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000448773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000448773.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000454690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000454690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000454690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000454690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000454690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
ENST00000585007.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200007195.hg.1 |
NR_039922.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003179.hg.1 |
TC02000215.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003179.hg.1 |
ENST00000317907.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003179.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02003179.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200149652.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149652.hg.1 |
ENST00000438654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149652.hg.1 |
ENST00000454690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149652.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200149652.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200149652.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149652.hg.1 |
ENST00000438654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149652.hg.1 |
ENST00000454690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149652.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200149652.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200149652.hg.1 |
TC0200007195.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000317907.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000428527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000428527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000428527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000428527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000428527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000428527.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000433416.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000433416.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000433416.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000433416.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000433416.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000433416.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000438654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000438654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000438654.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000448773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000448773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000448773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000448773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000448773.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000454690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000454690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000454690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000454690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000454690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
ENST00000585007.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_001193509.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NM_017735.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200007195.hg.1 |
NR_039922.1 |
Refseq |