Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015030.1 | NP_055845.1 | 27 - 28 | 3599 | 126 | |
XM_005248090.4 | XP_005248147.1 | 28 - 29 | 3657 | 126 | ||
XM_005248093.4 | XP_005248150.1 | 2 - 3 | 284 | 126 | ||
XM_011513680.2 | XP_011511982.1 | 25 - 26 | 3119 | 126 | ||
XM_011513681.2 | XP_011511983.1 | 25 - 26 | 3119 | 126 | ||
XM_011513682.2 | XP_011511984.1 | 24 - 25 | 3074 | 126 | ||
XM_011513683.2 | XP_011511985.1 | 27 - 28 | 3493 | 126 | ||
XM_011513684.2 | XP_011511986.1 | 2 - 3 | 290 | 126 | ||
XM_011513685.2 | XP_011511987.1 | 25 - 26 | 3119 | 126 | ||
XM_017008046.1 | XP_016863535.1 | 28 - 29 | 3924 | 126 | ||
GenBank mRNA | CR627228.1 | - | 11 - 12 | 1645 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |