Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023986.hg.1 |
TC06001948.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023986.hg.1 |
ENST00000369320.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023986.hg.1 |
ENST00000369325.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023986.hg.1 |
ENST00000369327.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023986.hg.1 |
ENST00000369329.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023986.hg.1 |
ENST00000369332.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023986.hg.1 |
ENST00000450832.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023986.hg.1 |
ENST00000479630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023986.hg.1 |
NM_003188.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023986.hg.1 |
NM_145331.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023986.hg.1 |
NM_145332.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023986.hg.1 |
NM_145333.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186669.hg.1 |
ENST00000369320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186669.hg.1 |
ENST00000369329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186669.hg.1 |
ENST00000369332.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186669.hg.1 |
ENST00000479630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186669.hg.1 |
NM_003188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186669.hg.1 |
NM_145331.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186669.hg.1 |
ENST00000369320.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186669.hg.1 |
ENST00000369329.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186669.hg.1 |
ENST00000369332.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186669.hg.1 |
ENST00000479630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186669.hg.1 |
NM_003188.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186669.hg.1 |
NM_145331.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186669.hg.1 |
TC0600012566.hg.1 |
Clariom D TC |