Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193451.1 | NP_001180380.1 | 18 | 8624 | 82 | |
NM_175861.3 | NP_787057.2 | 18 | 8715 | 82 | ||
XM_005253499.4 | XP_005253556.1 | 20 | 8876 | 82 | ||
XM_017020003.1 | XP_016875492.1 | 19 | 8709 | 82 | ||
XM_017020004.1 | XP_016875493.1 | 19 | 8669 | 82 | ||
XM_017020005.1 | XP_016875494.1 | 17 | 8482 | 82 | ||
XM_017020007.1 | XP_016875496.1 | 16 | 7902 | 82 | ||
GenBank mRNA | BC017984.1 | - | 1 | 1067 | 82 |
Set Membership: |
Detect Genomic DNA Both primers and probe map within a single exon |