Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012383.4 | NP_036515.4 | 7 - 8 | 619 | 228 | |
XM_006717053.3 | XP_006717116.1 | 7 - 8 | 628 | 228 | ||
XM_006717055.3 | XP_006717118.1 | 6 - 7 | 571 | 228 | ||
XM_011518524.2 | XP_011516826.1 | 8 - 9 | 438 | 228 | ||
XM_011518525.2 | XP_011516827.1 | 7 - 8 | 630 | 228 | ||
XM_017014622.1 | XP_016870111.1 | 7 - 8 | 620 | 228 | ||
GenBank mRNA | AK222596.1 | - | 7 - 8 | 563 | 228 | |
AK291511.1 | - | 7 - 8 | 587 | 228 | ||
BC007459.1 | - | 7 - 8 | 525 | 4052054 | 228 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |