Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000587637.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000587637.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000587637.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000587637.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000587637.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000590034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000590034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000590034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000590034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000590034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000590034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000590034.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000785.hg.1 |
ENST00000221494.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000785.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19000785.hg.1 |
TC19000050.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1900145761.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145761.hg.1 |
ENST00000587637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145761.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145761.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900145761.hg.1 |
TC1900006578.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900145761.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145761.hg.1 |
ENST00000587637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145761.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900145761.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000221494.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000586396.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000587637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000587637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000587637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000587637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000587637.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000589118.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000590034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000590034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000590034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000590034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000590034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000590034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000590034.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592314.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
ENST00000592839.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900006578.hg.1 |
NM_007165.4 |
Refseq |