Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK090520.1 | - | 2 - 3 | 300 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Homo sapiens Probe spans exons |