Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023207.hg.1 |
ENST00000221399.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023207.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19023207.hg.1 |
TC19001701.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000520977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000520977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000520977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000520977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000520977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000520977.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000522229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000522229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000522229.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000522341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000522341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000522341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000522341.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000520977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000520977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000520977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000520977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000520977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000520977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000522229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000522229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000522229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000522341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000522341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000522341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000522341.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900011103.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192938.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192938.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192938.hg.1 |
ENST00000520977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192938.hg.1 |
ENST00000522229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192938.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192938.hg.1 |
NM_003323.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900192938.hg.1 |
TC1900011103.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900192938.hg.1 |
ENST00000221399.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192938.hg.1 |
ENST00000518572.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192938.hg.1 |
ENST00000520977.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192938.hg.1 |
ENST00000522229.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192938.hg.1 |
ENST00000522945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900192938.hg.1 |
NM_003323.2 |
Refseq |