Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAZ1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032408.3 | NP_115784.1 | 18 - 19 | 4450 | 96 | |
XM_017012773.1 | XP_016868262.1 | 18 - 19 | 4463 | 96 | ||
GenBank mRNA | AB032253.1 | - | 18 - 19 | 4443 | 96 | |
AF072810.1 | - | 18 - 19 | 4450 | 96 | ||
AF084479.1 | - | 18 - 19 | 4382 | 96 | ||
BC136520.1 | - | 18 - 19 | 4283 | 9020510 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |