Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000430982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000430982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000430982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000430982.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000437572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000437572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000437572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000437572.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000451138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000451138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
ENST00000451138.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000419981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000419981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000419981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000419981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000419981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000419981.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000430029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000430029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000430029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000430029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000430029.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000485746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000485746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000485746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000485746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
ENST00000485746.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025055.hg.1 |
TC07003365.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025055.hg.1 |
ENST00000292478.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025055.hg.1 |
ENST00000438524.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07025055.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000430982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000430982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000430982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000430982.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000437572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000437572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000437572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000437572.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000451138.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000451138.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
ENST00000451138.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013592.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000419981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000419981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000419981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000419981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000419981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000419981.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000430029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000430029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000430029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000430029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000430029.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000485746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000485746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000485746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000485746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
ENST00000485746.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700013590.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201234.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201234.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201234.hg.1 |
ENST00000292478.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201234.hg.1 |
NM_015545.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201234.hg.1 |
TC0700013590.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700201286.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201286.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201286.hg.1 |
ENST00000413834.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700201286.hg.1 |
NM_001198879.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700201286.hg.1 |
TC0700013592.hg.1 |
Clariom D TC |