Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000480576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000480576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000480576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000480576.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000484989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000484989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000484989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
ENST00000484989.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005553.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005553.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005553.hg.1 |
TC13000491.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000480576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000480576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000480576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000480576.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000484989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000484989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000484989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
ENST00000484989.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008359.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156425.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156425.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156425.hg.1 |
ENST00000381989.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156425.hg.1 |
NM_006437.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156425.hg.1 |
TC1300008359.hg.1 |
Clariom D TC |