Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017266.hg.1 |
TC04001267.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017266.hg.1 |
ENST00000310613.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017266.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017272.hg.1 |
TC04001267.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017272.hg.1 |
ENST00000310613.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017272.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017268.hg.1 |
TC04001267.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017268.hg.1 |
ENST00000310613.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017268.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017265.hg.1 |
TC04001267.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017265.hg.1 |
ENST00000310613.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017265.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017264.hg.1 |
TC04001267.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017264.hg.1 |
ENST00000310613.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR04017264.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174403.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174403.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174403.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174403.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174403.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174403.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174403.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174403.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174403.hg.1 |
TC0400010940.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400174405.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174405.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174405.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174405.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174405.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174405.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174405.hg.1 |
TC0400010940.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400174407.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174407.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174407.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174407.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174407.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174407.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174407.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174407.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174407.hg.1 |
TC0400010940.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400174402.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174402.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174402.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174402.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174402.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174402.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174402.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174402.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174402.hg.1 |
TC0400010940.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0400174408.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174408.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174408.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174408.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174408.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174408.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174408.hg.1 |
TC0400010940.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000510821.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
ENST00000512870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0400010940.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174409.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174409.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174409.hg.1 |
ENST00000310613.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0400174409.hg.1 |
NM_014465.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0400174409.hg.1 |
TC0400010940.hg.1 |
Clariom D TC |