Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARID1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020732.3 | NP_065783.3 | 3 - 4 | 1780 | 66 | |
XM_011535984.2 | XP_011534286.2 | 3 - 4 | 1856 | 66 | ||
XM_017011103.1 | XP_016866592.1 | 3 - 4 | 1856 | 66 | ||
XM_017011104.1 | XP_016866593.1 | 3 - 4 | 1855 | 66 | ||
XM_017011105.1 | XP_016866594.1 | 3 - 4 | 1856 | 66 | ||
XM_017011106.1 | XP_016866595.1 | 3 - 4 | 1855 | 66 | ||
XM_017011107.1 | XP_016866596.1 | 3 - 4 | 1856 | 66 | ||
XM_017011108.1 | XP_016866597.1 | 3 - 4 | 1863 | 66 | ||
GenBank mRNA | AF468300.1 | - | 3 - 4 | 310 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Probe spans exons |