Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047455.hg.1 |
TC01003044.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047455.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047455.hg.1 |
ENST00000369426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047455.hg.1 |
ENST00000537870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047455.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047455.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047461.hg.1 |
TC01003044.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047461.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047461.hg.1 |
ENST00000369426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047461.hg.1 |
ENST00000497761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047461.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047461.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000497402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000497402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000497402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000497402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000497402.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000497761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
ENST00000497761.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047454.hg.1 |
TC01003044.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047454.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047454.hg.1 |
ENST00000369426.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047454.hg.1 |
ENST00000537870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047454.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01047454.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000497402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000497402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000497402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000497402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000497402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000497761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
ENST00000497761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018471.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242716.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242716.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242716.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242716.hg.1 |
ENST00000497761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242716.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242716.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242716.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242716.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242716.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242716.hg.1 |
ENST00000497761.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242716.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242716.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242716.hg.1 |
TC0100018471.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100242713.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242713.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242713.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242713.hg.1 |
ENST00000497402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242713.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242713.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242713.hg.1 |
ENST00000235521.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242713.hg.1 |
ENST00000369426.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242713.hg.1 |
ENST00000495746.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242713.hg.1 |
ENST00000497402.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242713.hg.1 |
NM_015836.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242713.hg.1 |
NM_201263.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242713.hg.1 |
TC0100018471.hg.1 |
Clariom D TC |