Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010792.hg.1 |
ENST00000553575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010792.hg.1 |
ENST00000553575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010792.hg.1 |
ENST00000556120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010792.hg.1 |
ENST00000556120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010792.hg.1 |
ENST00000556120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010792.hg.1 |
ENST00000556120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010792.hg.1 |
ENST00000556120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010792.hg.1 |
ENST00000556120.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010792.hg.1 |
NR_002770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010792.hg.1 |
NR_002770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010792.hg.1 |
NR_002770.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1400010792.hg.1 |
NR_002770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010792.hg.1 |
ENST00000553575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010792.hg.1 |
ENST00000553575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010792.hg.1 |
ENST00000556120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010792.hg.1 |
ENST00000556120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010792.hg.1 |
ENST00000556120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010792.hg.1 |
ENST00000556120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010792.hg.1 |
ENST00000556120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010792.hg.1 |
ENST00000556120.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1400010792.hg.1 |
NR_002770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010792.hg.1 |
NR_002770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010792.hg.1 |
NR_002770.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1400010792.hg.1 |
NR_002770.1 |
Refseq |