Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
ENST00000333447.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
ENST00000407050.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
ENST00000443686.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
ENST00000541501.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
ENST00000548586.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
ENST00000549325.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
ENST00000549396.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
ENST00000552948.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
NM_001220473.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
NM_001220474.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
NM_001220475.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
NM_001220476.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
NM_001220477.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
NM_001220478.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
NM_003625.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12023426.hg.1 |
TC12001776.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000333447.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000407050.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000443686.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000548586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000549325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000549396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000550359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000550584.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000551147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000552948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_001220473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_001220474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_001220475.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_001220476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_001220477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_001220478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_001282536.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_003625.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000333447.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000407050.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000443686.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000548586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000549325.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000549396.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000550359.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000550584.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000551147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
ENST00000552948.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_001220473.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_001220474.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_001220475.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_001220476.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_001220477.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_001220478.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_001282536.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
NM_003625.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200188227.hg.1 |
TC1200011393.hg.1 |
Clariom D TC |