Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZWILCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287821.1 | NP_001274750.1 | 2 - 3 | 453 | 208 | |
NM_017975.4 | NP_060445.3 | 2 - 3 | 358 | 113 | ||
GenBank mRNA | AK023175.1 | - | 2 - 3 | 348 | 113 | |
BC036900.1 | - | 2 - 3 | 104 | 5535106 | 113 | |
BC090041.1 | - | 2 - 3 | 202 | 6472990 | 208 | |
BX640701.1 | - | 2 - 3 | 420 | 208 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons |